Fumehoods
Our fume hoods meet the requirements of industry standards such as SEFA and ASHRAE. All our fume hoods are accompanied with individual typical test reports with commissioning on site.
Each of our hood type and size is factory tested to attain industry recommended containment level of less than 0.05 ppm. Our fume hoods can be configured with flammable, acid, vacuum or general base storage cabinets. Our flammable base cabinets conform to NFPA 30 requirements. They are also comply with FM approval class 6050 and UL 1275.
Hamilton Concept ® Laboratory Fume Hood
The fume hood are UL 1805 Classified, features the UL 1805 label on the hood face and have been tested to the ASHRAE 110 Standard with a containment rating of 4.0AM 0.05PPM. The baffles are fixed to maximize the capture and airflow inside the fume hood while minimizing static pressure and noise levels. The sash controlled by a single counterbalance, connected with a chain link system. This system allows for adjustment. The sight line for the hood is 35" high. Sash lowers automatically and stops at the 18" opening when released from full-open. A toggle lock is installed to secure the sash at the full-open position while loading or unloading the fume hood. The sash is available as a frameless vertical rising sash or a top hung combination sash. The top hung combo sash utilizes a narrow frame and bottom rail to offer more viewing area and ease of access. The sash airfoil has a low profile to facilitate ease of access to the worksurface with room for electrical cords to fit beneath it.
These hoods are available in: 4, 5, 6, 7, and 8-foot widths, and depths of 31.25", 37.24" and 43.25".