Products
Enabling the science that shapes the future
Visit us at booth #2118 to see how you can improve scientific outcomes, optimize operational productivity and advance clinical capabilities. You will also be able to view demonstrations of our new technologies and testing solutions and talk with our staff to see how we can help meet your needs.
Thermo Scientific B·R·A·H·M·S PlGF plus KRYPTOR and Thermo Scientific B·R·A·H·M·S sFlt-1 KRYPTOR Assays
Thermo Scientific™ B·R·A·H·M·S™ PlGF plus KRYPTOR™ and Thermo Scientific™ B·R·A·H·M·S™ sFlt-1 KRYPTOR™ Assays are used in conjunction with B·R·A·H·M·S sFlt-1 KRYPTOR assays (as well as other laboratory tests and clinical assessments) to help assess pregnant women who are hospitalized with hypertensive disorders and their risk of preeclampsia with severe features, all within two weeks of presentation.
- First and only immunoassays to receive breakthrough designation and FDA clearance for the risk assessment and clinical management of preeclampsia
Thermo Scientific Indiko and Indiko Plus Analyzers
Thermo Scientific™ Indiko™ and Thermo Scientific™ Indiko Plus™ Clinical and Specialty Chemistry Analyzers are easy-to-use, cost-effective systems designed for load-up and walk-away convenience. An intuitive user interface and many automated features help you manage your daily workflow. These self-contained analyzers have small footprints and fit well in laboratories with space limitations.
Drugs of Abuse Assays
Thermo Scientific drugs of abuse tests use well-established DRI™ and CEDIA™ technologies, which have proven assay performance, are easy to use, and are valued for accuracy, specificity, and reproducibility.
- High concordance with confirmatory
methods: 95% - Excellent inter-assay and between-run assay precision: <10% CV
- Applications available on a wide range of commonly used chemistry analyzers
- Minimal retesting optimizes efficiency while minimizing operating costs
Thermo Scientific Quality Control and Assurance Program
The Thermo Scientific™ MAS™ Quality Control (QC) portfolio offers improved efficiency with stable and consolidated third party solutions to support QC management. It delivers accurate results to help you ensure reliable patient results.
- Simplify QC management processes through consolidation of controls
- Optimize product use with industry leading three-to-five year shelf life and a 30-day open vial stability*
- Save time and costs for your lab during your QC transition with our personalized EZ Switch program
*Refer to package insert for specific analyte and stability claims. Shelf life starts from the date of manufacture.
Thermo Scientific AcroMetrix Quality Controls and Standards
The Thermo Scientific™ AcroMetrix™ (AMX) portfolio offers standardized third-party controls, validation panels and custom solutions that will help you achieve accurate and consistent test results.
Thermo Scientific Phadia 250 Laboratory System
The Thermo Scientific™ Phadia™ 250 Laboratory System was designed to aid in the diagnosis of autoimmune diseases and IgE mediated allergic disorders, and provide true walk-away, automated testing on a single instrument. The assay menu for autoimmune diseases includes celiac, rheumatoid arthritis and connective tissue diseases, among others. The system provides quality and consistency with decades of market leadership and innovative recombinant protein technology and connects to Laboratory Automation Systems (LAS).
LC-MS/MS Medical Devices for In Vitro Diagnostic (IVD) Use*
The Thermo Scientific™ Quantis™ MD and Thermo Scientific™ Altis™ MD Systems offer comprehensive and reliable mass spectrometry solutions, enabling in vitro diagnostic analysis. Powered by a complete suite of software conforming to IVD requirements, with an optional laboratory information system (LIS) connection, it provides consistent, reliable results you can have confidence in. Couple this system with our Thermo Scientific™ Vanquish™ MD Liquid Chromatography Platform for the optimal LC-MS solution.
*For in vitro diagnostic use. Specifications subject to change. Availability of product in each country depends on local regulatory marketing authorization status.
Thermo Scientific Transcend DSX-1 UHPLC System*
Redefine your traditional, labor-intensive, dried matrix spot analysis workflow with the Thermo Scientific™ Transcend™ DSX-1 UHPLC System. This fully automated system for toxicology and anti-doping laboratories enables automation and efficient online extraction and enrichment, delivering sample compliance and minimal ambiguity for high throughput LC-MS/MS analysis in a single integrated system.
*General laboratory use. For toxicology use only.
Thermo Scientific Tox Explorer Collection
Navigate with ease—an all-in-one LC-MS/MS solution for toxicology. The Thermo Scientific™ Tox Explorer™ collection helps the world's toxicology community solve complex analytical challenges and increase laboratory productivity. Built on a single software solution, the collection comes equipped with a standardized LC-MS method with retention times, a (U)HPLC column, an experimentally obtained compound database and library, as well as on-site LC-MS/MS instrument setup and Tox Explorer application training, enabling smooth start-up and implementation.
For increased confidence in data and productivity in your laboratory, the collection is now available with Thermo Scientific™ Orbitrap™ MS and Thermo Scientific TSQ Plus triple quadrupole mass spectrometer platform options.
Applied Biosystems QuantStudio 5 Dx Real-Time PCR System
Designed to simplify workflows and minimize training needs, the Applied Biosystems QuantStudio 5 Dx Real-Time PCR System can help you get to your clinical answers quickly by fitting seamlessly into your established workflow. This compact, flexible system provides confidence in performance and supports both development and IVD modes.
Applied Biosystems QuantStudio 7 Pro Dx Real-Time PCR System
Powered by proven technology, the Applied Biosystems QuantStudio 7 Pro Dx Real-Time PCR System is a standalone qPCR platform that is versatile, highly expandable, and easy to use. Designed to support in vitro diagnostic (IVD) applications, the QuantStudio 7 Pro Dx system incorporates smart features such as hand-free functionality. Whether your laboratory is running routine assays or developing new ones, the QuantStudio 7 Pro Dx can support your work.
Applied Biosystems Diomni Enterprise Software
Applied Biosystems™ Diomni™ Enterprise Software is an intelligent, on-premise workflow solution that connects all steps in your real-time PCR (qPCR) test development and routine testing workflows with one single ecosystem. Experience the power of one: one connection, one interface, one workflow.
HIV-1 Genotyping Kit with Integrase*
The Applied Biosystems HIV-1 Genotyping Kit with integrase is designed to genotype the protease, reverse transcriptase (RT), and integrase (IN) regions of the HIV-1 pol gene. The genotyping result encompasses codons 6-99 in the PR region, codons 1-251 in the RT region, and codons 1-288 of the IN region of the gene. HIV-1 genomic mutations in these regions confer resistance to anti-retroviral drugs that act through the inhibition of the protease and reverse transcriptase.
*For research use only. Not for use in diagnostic procedures.
Applied Biosystems TaqMan PGx Express Panel*
This Applied Biosystems TaqMan PGx (Pharmacogenomics) Express Panel is offered in pre-spotted 384-well plates, an efficient, easy-to-use format for pharmacogenomics applications. The express panel contains 64 TaqMan DME and SNP genotyping assays per 384-well plate. The assays were developed to detect polymorphisms in genes encoding drug metabolism enzymes (DMEs) and associated transport proteins. This panel provides coverage of essential, commonly studied markers within CYP2D6, CYP2C9, CYP2C19, and other important DME and clinical research genes.
*For research use only. Not for use in diagnostic procedures.
Applied Biosystems TrueMark STI Select Panel Combo Kit*
The Applied Biosystems™ TrueMark™ STI Select Panel Combo Kit is a multiplex real‐time PCR solution that contains the assay and master mix needed for simultaneous detection of RNA from Chlamydia trachomatis (CT), Neisseria gonorrhoeae (NG), Mycoplasma genitalium (MG), and Trichomonas vaginalis (TV) in extracted human samples. Human ribonuclease P (RNase P) is included as an endogenous sample collection control.
*For research use only. Not for use in diagnostic procedures.