Which Centrifuge Is Right for You?
Answer the following multiple-choice questions and we’ll suggest a centrifuge to fit your needs.
What are you spinning?
Will the centrifuge be used for STAT, routine, or PRP processing?
Would you prefer a fully programmable digital unit or three set-and-lock presets?
Where will your centrifuge be used?
Do you prefer single speed or multiple/programmable options?
Would you prefer a fully programmable digital unit or three set-and-lock presets?
Fisherbrand DASH Flex 12 and Fisherbrand 12 Centrifuges
Unique flashing lid lights make it easy to see when your Fisherbrand™ 12 or DASH Flex 12 centrifuge has finished its cycle. Choose a digital display or a two-button interface — both have presets for common spin times and speeds, and they allow you to add your own validated program.
Shop Now
Fisherbrand DASH Flex 12 and Fisherbrand 12 Centrifuges
Unique flashing lid lights make it easy to see when your Fisherbrand™ 12 or DASH Flex 12 centrifuge has finished its cycle. Choose a digital display or a two-button interface — both have presets for common spin times and speeds, and they allow you to add your own validated program.
Shop Now
Fisherbrand Model 642M Centrifuges
Designed for mobility, the Fisherbrand Model 642M Centrifuge is powered by a vehicle’s 12V electrical supply. It features a single pre-set runtime and speed — 10 minutes at 3400rpm/1600 x g — to spin common chemistry tubes.
Shop Now
Fisherbrand Model 642E Centrifuges
The easy two-button operation of Fisherbrand Model 642E Centrifuges ensures repeatability and standardized 10-minute spins at 1600 x g. Their small footprint and six-tube capacity work well in doctors’ offices, patient service centers, and other outreach locations.
Shop Now
Fisherbrand HORIZON Flex Centrifuges
Get easy access to all of your saved spin protocols with Fisherbrand HORIZON Flex Programmable Centrifuges. Customize settings using the digital display and save up to 10 different centrifuge cycles.
Shop Now
Fisherbrand Horizon 6 FA and 6 Flex FA Centrifuges
Process urine, PFP, and other samples with the fixed-angle rotors of Fisherbrand HORIZON 6 FA Centrifuges. The 6 Flex FA offers a digital display and 10 programmable cycles, and it lets you set and lock your settings for more consistent sample handling.
Shop Now