Fisher Safety
What's being discontinued?
All AV-3000 and AV-2000 facepieces with the black nosecup and the following head harnesses will be discontinued on Dec. 31, 2012.
If you are currently using an AV-3000 or AV-2000 Facepiece with a black nosecup ...
AV-3000Scott Safety AV-3000
  • Kevlar* (805773 series)
  • Polyester (805774 series)
  • Rubber (805775 series)
* Note: Scott Safety will make Kevlar versions of the AV-3000 with black nosecups available beyond 2012 for existing customers using Air-Pak Fifty SCBA labeled with a red approval sticker "NFPA 1981: 1997 Edition" in order to support NFPA approval.
AV-2000Scott Safety AV-2000
  • Kevlar* (804191 series)
  • Polyester (804069 series)
  • Rubber (805257 series)
* Note: Scott Safety will make Kevlar versions of the AV-3000 with black nosecups available beyond 2012 for existing customers using Air-Pak Fifty SCBA labeled with a red approval sticker “NFPA 1981: 1997 Edition” in order to support NFPA approval.
Customer Options
Upgrade to new AV-3000 Series facepiece
If you are currently using the AV-3000 facepiece with the black nosecup, Scott Safety has several newer facepiece options to select from to replace your existing facepiece. The AV-3000 with SureSeal system is the most popular Scott Safety facepiece sold today—offering greater comfort and better fit, plus a whole array of other features.
See all Scott AV-3000 Facepieces with SureSeal »
See all Scott AV-3000 Facepieces »
See the AV-3000 conversion table »
See the AV-2000 conversion table »
Continued use of your existing facepiece
If you are currently using an AV-3000 or AV-2000 facepiece with a black nosecup, you can continue using it. The respiratory approvals will remain in effect. Should you need to replace the black nosecup, the cup will be available through Dec. 31, 2012. Please note: the AV-3000 and AV-2000 facepieces (with black nosecup) will no longer be available for sale after Dec. 31, 2012.
Upgrade the black nosecup to the grey nosecup
AV-3000 facepieces currently outfitted with black nosecups can be upgraded to grey nosecups by simply purchasing corresponding nosecup; no other conversion kit is required. AV-2000 facepieces currently outfitted with black nosecups can be upgraded to grey nosecups by simply purchasing the upgrade kit for those facepieces that are upgradable. More information »